accessibility menu, dialog, popup

UserName

OHHNOS

CAS RN 98902-53-5

IUPAC Name: N-(4-hydroxyphenyl)thiophene-2-carboxamide
Synonyms: N-(4-hydroxyphenyl)thiophene-2-carboxamide
Molecular Weight (g/mol): 219.26
Molecular Formula: C11H9NO2S
InChi Key: XBRSNMYZHZKDSW-UHFFFAOYSA-N
SMILES: OC1=CC=C(NC(=O)C2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results