CAS RN 98603-84-0
CAS RN 98603-84-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sialyl Lewis X, TRC
CAS: 98603-84-0 Molecular Formula: C31 H52 N2 O23 Molecular Weight (g/mol): 820.74 IUPAC Name: (2S,4S,5R,6R)-5-acetamido-2-[(2S,3R,4S,5S,6R)-2-[(2R,3R,4R,5R)-5-acetamido-1,2-dihydroxy-6-oxo-4-[(2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyhexan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid SMILES: C[C@@H]1O[C@@H](O[C@H]([C@@H](NC(=O)C)C=O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O[C@@]3(C[C@H](O)[C@@H](NC(=O)C)[C@@H](O3)[C@H](O)[C@H](O)CO)C(=O)O)[C@H]2O)[C@H](O)CO)[C@@H](O)[C@H](O)[C@@H]1O