accessibility menu, dialog, popup

UserName

CAS RN 97-14-3

H3COHNOHOHO

CAS RN 97-14-3

IUPAC Name: 2-acetamido-3-hydroxypropanoic acid
Synonyms: 2-acetamido-3-hydroxypropanoic acid
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: JJIHLJJYMXLCOY-UHFFFAOYNA-N
SMILES: CC(=O)NC(CO)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)