accessibility menu, dialog, popup

UserName

CAS RN 96449-68-2

NOFOHO

CAS RN 96449-68-2

IUPAC Name: 1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid
Synonyms: 1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 237.23
Molecular Formula: C12H12FNO3
InChi Key: IYAFJFDPMVQTJO-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CC2=CC=C(F)C=C2)C(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results