accessibility menu, dialog, popup

UserName

H3COSOOOCH3

CAS RN 95735-63-0

IUPAC Name: 2-[(3,4-dimethoxyphenyl)sulfanyl]acetate
Synonyms: 2-[(3,4-dimethoxyphenyl)sulfanyl]acetate
Molecular Weight (g/mol): 227.25
Molecular Formula: C10H11O4S
InChi Key: ANVXBTSSFGHHBD-UHFFFAOYSA-M
SMILES: COC1=CC=C(SCC([O-])=O)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results