accessibility menu, dialog, popup

UserName

CAS RN 95211-71-5

CH3SOHNN

CAS RN 95211-71-5

IUPAC Name: 11-methyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),5-trien-3-one
Synonyms: 11-methyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),5-trien-3-one
Molecular Weight (g/mol): 220.29
Molecular Formula: C11H12N2OS
InChi Key: YSMXTYUERRJGMR-UHFFFAOYNA-N
SMILES: CC1CCC2=C(C1)SC1=C2C(=O)NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results