accessibility menu, dialog, popup

UserName

CAS RN 949902-02-7

NOSOOFFF

CAS RN 949902-02-7

IUPAC Name: 8-benzyl-8-azabicyclo[3.2.1]oct-2-en-3-yl trifluoromethanesulfonate
Synonyms: 8-benzyl-8-azabicyclo[3.2.1]oct-2-en-3-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 347.35
Molecular Formula: C15H16F3NO3S
InChi Key: GBUXWXWPNDHMRG-UHFFFAOYNA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC2CCC(C1)N2CC1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results