accessibility menu, dialog, popup

UserName

CAS RN 942206-18-0

IUPAC Name: 3,6-dibromo-2-chloropyridine
Synonyms: 3,6-dibromo-2-chloropyridine
Molecular Weight (g/mol): 271.34
Molecular Formula: C5H2Br2ClN
InChi Key: CVZSNXGMWIGPBF-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=CC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results