accessibility menu, dialog, popup

UserName

CAS RN 939793-16-5

CH3H3CCH3OONBr

CAS RN 939793-16-5

IUPAC Name: tert-butyl 3-bromopyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-bromopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 250.14
Molecular Formula: C9H16BrNO2
InChi Key: QJTKPXFJOXKUEY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(Br)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results