CAS RN 93813-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
16-Heptadecynoic Acid, TRC
CAS: 93813-16-2 Molecular Formula: C17H30O2 Molecular Weight (g/mol): 266.42 IUPAC Name: heptadec-16-ynoic acid SMILES: OC(=O)CCCCCCCCCCCCCCC#C