accessibility menu, dialog, popup

UserName

NHOBrCl

CAS RN 934185-00-9

IUPAC Name: 2-bromo-N-[(4-chlorophenyl)methyl]acetamide
Synonyms: 2-bromo-N-[(4-chlorophenyl)methyl]acetamide
Molecular Weight (g/mol): 262.53
Molecular Formula: C9H9BrClNO
InChi Key: HFWRJHHLKNQBHJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CNC(=O)CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results