accessibility menu, dialog, popup

UserName

CAS RN 93391-29-8

LiH3CH3COO

CAS RN 93391-29-8

IUPAC Name: lithium(1+) 2-propylpentanoate
Synonyms: lithium(1+) valproate
Molecular Weight (g/mol): 150.15
Molecular Formula: C8H15LiO2
InChi Key: CWFAFDCTBAYTFA-UHFFFAOYSA-M
SMILES: [Li+].CCCC(CCC)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results