CAS RN 933-78-8
CAS RN 933-78-8
IUPAC Name:
2,3,5-trichlorophenol
Synonyms:
2,3,5-trichlorophenol
Molecular Weight (g/mol):
197.44
Molecular Formula:
C6H3Cl3O
InChi Key:
WWGQHTJIFOQAOC-UHFFFAOYSA-N
SMILES:
OC1=CC(Cl)=CC(Cl)=C1Cl
1
–
2
of
2
results
1
2,3,5-Trichlorophenol 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality