accessibility menu, dialog, popup

UserName

CAS RN 932-31-0

MgBrCH3

CAS RN 932-31-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 195.34
Molecular Formula: C7H7BrMg
InChi Key: YAMQOOCGNXAQGW-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].Cc1cccc[cH-]1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results