accessibility menu, dialog, popup

UserName

H3CPOClCl

CAS RN 928-64-3

IUPAC Name: hexylphosphonoyl dichloride
Synonyms: hexylphosphonoyl dichloride
Molecular Weight (g/mol): 203.04
Molecular Formula: C6H13Cl2OP
InChi Key: GZXBQDURAKBNAZ-UHFFFAOYSA-N
SMILES: CCCCCCP(Cl)(Cl)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results