CAS RN 92692-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amorphadiene, TRC
CAS: 92692-39-2 Molecular Formula: C15 H24 Molecular Weight (g/mol): 204.35 Synonym: Naphthalene, 1,2,3,4,4a,5,6,8a-octahydro-4,7-dimethyl-1-(1-methylethenyl)-, (1R,4R,4aS,8aR)-,Naphthalene, 1,2,3,4,4a,5,6,8a-octahydro-4,7-dimethyl-1-(1-methylethenyl)-, [1R-(1α,4β,4aβ,8aβ)]-,(1R,4R,4aS,8aR)-1,2,3,4,4a,5,6,8a-Octahydro-4,7-dimethyl-1-(1-methylethenyl)naphthalene,(-)-Amorpha-4,11-diene,Amorpha-4,11-diene,Amorphadiene IUPAC Name: (1R,4R,4aS,8aR)-4,7-dimethyl-1-prop-1-en-2-yl-1,2,3,4,4a,5,6,8a-octahydronaphthalene SMILES: C[C@@H]1CC[C@H]([C@H]2C=C(C)CC[C@@H]12)C(=C)C