accessibility menu, dialog, popup

UserName

CAS RN 92636-38-9

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]-1H-pyrrole
Synonyms: 1-[4-(trifluoromethyl)phenyl]pyrrole
Molecular Weight (g/mol): 211.19
Molecular Formula: C11H8F3N
InChi Key: JFGOCJGHXODFKU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)N1C=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results