CAS RN 92471-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,2S,4R)-2-Hydroxy-Alpha,Alpha,4-trimethylcyclohexanemethanol, TRC
CAS: 92471-23-3 Molecular Formula: C10 H20 O2 Molecular Weight (g/mol): 172.26 Synonym: [1R-(1alpha,2alpha,4beta)]-2-hydroxy-alpha,alpha,4-trimethylcyclohexanemethanol SMILES: C[C@@H]1CC[C@H]([C@@H](O)C1)C(C)(C)O