accessibility menu, dialog, popup

UserName

H3COFClCl

CAS RN 916420-72-9

IUPAC Name: 1-(3,6-dichloro-2-fluorophenyl)ethan-1-one
Synonyms: 1-(3,6-dichloro-2-fluorophenyl)ethanone
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H5Cl2FO
InChi Key: KIPVBHCZTSPODV-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results