accessibility menu, dialog, popup

UserName

CAS RN 913836-15-4

IUPAC Name: (6-chloro-2-methylpyridin-3-yl)boronic acid
Synonyms: 6-chloro-2-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 171.39
Molecular Formula: C6H7BClNO2
InChi Key: WEKRUIAZOLIAOG-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Cl)=N1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results