CAS RN 91271-83-9
IUPAC Name:
4-{[3-(azaniumylmethyl)phenyl]methyl}morpholin-4-ium
Synonyms:
4-{[3-(ammoniomethyl)phenyl]methyl}morpholin-4-ium
Molecular Weight (g/mol):
208.3
Molecular Formula:
C12H20N2O
InChi Key:
JCIFAZAIURDZQN-UHFFFAOYSA-P
SMILES:
[NH3+]CC1=CC=CC(C[NH+]2CCOCC2)=C1
1
–
1
of
1
results
1
[3-(Morpholinomethyl)phenyl]methylamine, 97%, Thermo Scientific™
CAS: 91271-83-9 Molecular Formula: C12H18N2O Molecular Weight (g/mol): 206.289 MDL Number: MFCD06657777 InChI Key: JCIFAZAIURDZQN-UHFFFAOYSA-N Synonym: 3-morpholin-4-ylmethyl-benzylamine,3-morpholinomethyl phenyl methylamine,3-morpholinomethyl phenyl methanamine,3-morpholin-4-ylmethyl phenyl methanamine,3-morpholin-4-yl methyl phenyl methanamine,1-3-morpholin-4-ylmethyl phenyl methanamine,3-morpholin-4-ylmethyl phenyl methylamine,3-morpholin-4-ylmethylbenzylamine,3-morpholin-4-ylmethyl benzylamine PubChem CID: 6484182 IUPAC Name: [3-(morpholin-4-ylmethyl)phenyl]methanamine SMILES: C1COCCN1CC2=CC(=CC=C2)CN