CAS RN 912671-55-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy Estradiol 6-N3-Adenine, TRC
CAS: 912671-55-7 Molecular Formula: C23H27N5O3 Molecular Weight (g/mol): 421.49 Synonym: (17β)-6-(6-Amino-3H-purin-3-yl)-estra-1,3,5(10)-triene-2,3,17-triol SMILES: OC1=C(O)C=C2C(C(N3C4=NC=NC4=C(N)N=C3)C[C@]5([H])[C@]2([H])CC[C@@]6(C)[C@@]5([H])CC[C@@H]6O)=C1