accessibility menu, dialog, popup

UserName

CAS RN 911295-24-4

CH3NHNOOHONHClHCl

CAS RN 911295-24-4

IUPAC Name: 4-[1-(4-hydroxyphenyl)-4-methyl-5-{4-[2-(piperidin-1-yl)ethoxy]phenyl}-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,5-dien-1-one dihydrochloride
Synonyms: methylpiperidinopyrazole
Molecular Weight (g/mol): 542.5
Molecular Formula: C29H33Cl2N3O3
InChi Key: SMPRQALBFACGGO-UHFFFAOYSA-N
SMILES: Cl.Cl.CC1=C(N(NC1=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1)C1=CC=C(OCCN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results