CAS RN 910299-74-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(11Beta,16Alpha)-11,16-Dihydroxyandrosta-1,4-diene-3,17-dione, TRC
CAS: 910299-74-0 Molecular Formula: C19H24O4 Molecular Weight (g/mol): 316.39 Synonym: Budesonide Related Compound IUPAC Name: (8S,9S,10R,11S,13S,14S,16R)-11,16-dihydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-dione SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)C2=O