accessibility menu, dialog, popup

UserName

CH3NNN

CAS RN 910036-91-8

IUPAC Name: 3-[(4-methyl-1,4-diazepan-1-yl)methyl]benzonitrile
Synonyms: 3-[(4-methyl-1,4-diazepan-1-yl)methyl]benzonitrile
Molecular Weight (g/mol): 229.33
Molecular Formula: C14H19N3
InChi Key: WCQJGGDNUFLTPA-UHFFFAOYSA-N
SMILES: CN1CCCN(CC2=CC=CC(=C2)C#N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results