CAS RN 91-55-4
CAS RN 91-55-4
IUPAC Name:
2,3-dimethyl-1H-indole
Synonyms:
1H-indole, 2,3-dimethyl-
Molecular Weight (g/mol):
145.21
Molecular Formula:
C10H11N
InChi Key:
PYFVEIDRTLBMHG-UHFFFAOYSA-N
SMILES:
CC1=C(C)C2=CC=CC=C2N1
1
–
1
of
1
results
1
2,3-Dimethylindole, 97%
CAS: 91-55-4 Molecular Formula: C10H11N Molecular Weight (g/mol): 145.205 MDL Number: MFCD00005617 InChI Key: PYFVEIDRTLBMHG-UHFFFAOYSA-N Synonym: 2,3-dimethylindole,1h-indole, 2,3-dimethyl,indole, 2,3-dimethyl,2,3-dimethyl indole,tfw7o9hwzk,unii-tfw7o9hwzk,2,3-dimethyl-1h-indol,2,3-dimethyl-indol,2,3-dimethyl-indole,pubchem7334 PubChem CID: 7053 IUPAC Name: 2,3-dimethyl-1H-indole SMILES: CC1=C(NC2=CC=CC=C12)C