CAS RN 90632-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Didechlorodihydroxycyclophosphamide (Technical Grade), TRC
CAS: 90632-33-0 Molecular Formula: C7H17N2O4P Molecular Weight (g/mol): 224.19 Synonym: 2-(Bis(2-hydroxyethyl)amino)-1,3,2-oxazaphosphinane 2-Oxide SMILES: OCCN(CCO)P1(=O)NCCCO1