CAS RN 90581-38-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(5R,6S)-6-[(1R)-1-Hydroxyethyl]-7-oxo-3-[(3R)-3-pyrrolidinylthio]-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic Acid (>90%), TRC
CAS: 90581-38-7 Molecular Formula: C13H18N2O4S Molecular Weight (g/mol): 298.36 Synonym: [5R-[3(R*),5alpha,6alpha(R*)]]-6-(1-Hydroxyethyl)-7-oxo-3-(3-pyrrolidinylthio)-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic Acid IUPAC Name: 6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(3S)-pyrrolidin-3-yl]sulfanyl-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid SMILES: C[C@@H](O)C1C2CC(=C(N2C1=O)C(=O)O)S[C@H]3CCNC3