CAS RN 9041-37-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sephadex LH-20, TRC
CAS: 9041-37-6 Molecular Formula: C9H8N2O3S Molecular Weight (g/mol): 224.23642 Synonym: SEPHADEX(R) LH-20,SEPHADEX LH-20,LIPOPHILIC SEPHADEX,DEXTRAN 20,sephadex(R) lh-20 for chromatography,SEPHADEX(R) LH-20 FOR CHROMATOGRAPHY, 25-100 UM*,Lipophilic Sephadex(R) IUPAC Name: methyl 1-acetyl-1H-thieno[3,2-c]pyrazole-5-carboxylate SMILES: CC(N1C2=C(SC(C(OC)=O)=C2)C=N1)=O