accessibility menu, dialog, popup

UserName

NHON

CAS RN 902837-34-7

IUPAC Name: 4-(piperidin-3-yloxy)benzonitrile
Synonyms: 4-(piperidin-3-yloxy)benzonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: KMBKWCFPFFJUPE-UHFFFAOYNA-N
SMILES: N#CC1=CC=C(OC2CCCNC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results