CAS RN 90-39-1
IUPAC Name:
(1S,2R,9S,10S)-7,15-diazatetracyclo[7.7.1.0²,⁷.0¹⁰,¹⁵]heptadecane
Synonyms:
sparteine l-sparteine 6β,7α,9α,11α-pachycarpine sparteine (inn) and its salts
Molecular Weight (g/mol):
234.39
Molecular Formula:
C15H26N2
InChi Key:
SLRCCWJSBJZJBV-ZQDZILKHSA-N
SMILES:
C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@H]34)[C@@H]2C1
1
–
2
of
2
results
2
(-)-Sparteine, MedChemExpress
MedChemExpress (-)-Sparteine is a natural alkaloid isolated from beans.