accessibility menu, dialog, popup

UserName

CAS RN 89641-18-9

H3CONBOHHOOH3CN

CAS RN 89641-18-9

IUPAC Name: (2,4-dimethoxypyrimidin-5-yl)boronic acid
Synonyms: 2,4-dimethoxypyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 183.96
Molecular Formula: C6H9BN2O4
InChi Key: LKGKUACPLXCVOF-UHFFFAOYSA-N
SMILES: COC1=NC=C(B(O)O)C(OC)=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)