CAS RN 895542-09-3
CAS RN 895542-09-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Trifarotene, MedChemExpress
MedChemExpress Trifarotene (CD5789) is a potent and selective RARγ agonist. Trifarotene (CD5789) shows ∼65-fold and ∼16-fold selectivitiy for the RARγ (EC50=7.7 nM) over RARα (EC50=500 nM) and RARβ (EC50=125 nM), respectively.