CAS RN 89363-50-8
IUPAC Name:
1-pentyl-4-(4-propylcyclohexyl)benzene
Synonyms:
1-pentyl-4-(4-propylcyclohexyl)benzene
Molecular Weight (g/mol):
272.48
Molecular Formula:
C20H32
InChi Key:
RSUADYMBPNVXJC-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=C(C=C1)C1CCC(CCC)CC1
1
–
1
of
1
results
1
trans-1-n-Pentyl-4-(4-n-propylcyclohexyl)benzene, 99%, Thermo Scientific™
CAS: 89363-50-8 Molecular Formula: C20H32 Molecular Weight (g/mol): 272.476 MDL Number: MFCD22125193 InChI Key: RSUADYMBPNVXJC-UHFFFAOYSA-N Synonym: 1-pentyl-4-trans-4-propylcyclohexyl benzene,1-4-propylcyclohexyl-4-pentylbenzene,4-trans-4-propylcyclohexyl pentylbenzene,4-trans-4-propylcyclohexyl-pentylbenzene,1-pentyl-4-4-propylcyclohexyl benzene,trans-1-n-pentyl-4-4-n-propylcyclohexyl benzene,1-pentyl-4-1s,4r-4-propylcyclohexyl benzene PubChem CID: 15606764 IUPAC Name: 1-pentyl-4-(4-propylcyclohexyl)benzene SMILES: CCCCCC1=CC=C(C=C1)C2CCC(CC2)CCC