CAS RN 890087-21-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
LUF6000, MedChemExpress
MedChemExpress LUF6000 is an orally active allosteric modulator of the A3 adenosine receptor. LUF6000 has potent anti-inflammatory effect.