accessibility menu, dialog, popup

UserName

CAS RN 887266-99-1

NFI

CAS RN 887266-99-1

IUPAC Name: 3-fluoro-4-iodobenzonitrile
Synonyms: 3-fluoro-4-iodobenzonitrile
Molecular Weight (g/mol): 247.01
Molecular Formula: C7H3FIN
InChi Key: NPKQMCCUPXZXFI-UHFFFAOYSA-N
SMILES: FC1=C(I)C=CC(=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results