accessibility menu, dialog, popup

UserName

CAS RN 886761-78-0

FFFFOH

CAS RN 886761-78-0

IUPAC Name: 2-[2-fluoro-4-(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 2-[2-fluoro-4-(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 208.16
Molecular Formula: C9H8F4O
InChi Key: ZZPFABXJRPFKOI-UHFFFAOYSA-N
SMILES: OCCC1=C(F)C=C(C=C1)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results