accessibility menu, dialog, popup

UserName

CH3OOHOOFFF

CAS RN 886502-37-0

IUPAC Name: 4-methoxy-2-(trifluoromethoxy)benzoic acid
Synonyms: 4-methoxy-2-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 236.15
Molecular Formula: C9H7F3O4
InChi Key: PDDVYCWKVQXOTR-UHFFFAOYSA-N
SMILES: COC1=CC(OC(F)(F)F)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results