accessibility menu, dialog, popup

UserName

CAS RN 886500-26-1

ClBrFFF

CAS RN 886500-26-1

IUPAC Name: 2-(bromomethyl)-1-chloro-3-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1-chloro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: FSQANOAIVKWIEU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(CBr)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results