accessibility menu, dialog, popup

UserName

OFFFHOF

CAS RN 886498-03-9

IUPAC Name: 2-fluoro-5-(trifluoromethoxy)phenol
Synonyms: 2-fluoro-5-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 196.1
Molecular Formula: C7H4F4O2
InChi Key: MKRRYWGEQVWJCA-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results