accessibility menu, dialog, popup

UserName

NNClI

CAS RN 885275-59-2

IUPAC Name: 6-chloro-3-iodoimidazo[1,2-a]pyridine
Synonyms: 6-chloro-3-iodoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 278.48
Molecular Formula: C7H4ClIN2
InChi Key: IHTZNIKMBKFWHV-UHFFFAOYSA-N
SMILES: ClC1=CN2C(I)=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results