CAS RN 885269-59-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Fluoro-1,2,3,4-tetrahydro-naphthalene-2-carboxylic Acid, TRC
CAS: 885269-59-0 Molecular Formula: C11 H11 F O2 Molecular Weight (g/mol): 194.2 IUPAC Name: 6-fluorotetralin-2-carboxylic acid SMILES: OC(=O)C1CCc2cc(F)ccc2C1