CAS RN 88460-72-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(5-formylfuran-2-yl)benzoic Acid, TRC
CAS: 88460-72-4 Molecular Formula: C12H8O4 Molecular Weight (g/mol): 216.18 SMILES: OC(=O)c1ccccc1c2oc(C=O)cc2