accessibility menu, dialog, popup

UserName

CH3H2NBOH3CH3CH3CCH3O

CAS RN 882670-69-1

IUPAC Name: 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: HPNBWHDRLCVZFV-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results