accessibility menu, dialog, popup

UserName

CAS RN 881189-62-4

IUPAC Name: 3-(3-fluoro-4-methylphenyl)propanoic acid
Synonyms: 3-(3-fluoro-4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H11FO2
InChi Key: DYVQAEHNFXNAEH-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results