accessibility menu, dialog, popup

UserName

CAS RN 874800-57-4

H3CONO

CAS RN 874800-57-4

IUPAC Name: 5-(2-methoxyphenyl)-1,2-oxazole
Synonyms: 5-(2-methoxyphenyl)-1,2-oxazole
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: DPPFYBGCJJBZEH-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CC=NO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results