accessibility menu, dialog, popup

UserName

CAS RN 871329-60-1

IUPAC Name: [3-(morpholine-4-sulfonyl)phenyl]boronic acid
Synonyms: 3-(morpholine-4-sulfonyl)phenylboronic acid
Molecular Weight (g/mol): 271.09
Molecular Formula: C10H14BNO5S
InChi Key: HIKVVOUXXNJGAK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results