accessibility menu, dialog, popup

UserName

H3CONHCH3H3C

CAS RN 868755-46-8

IUPAC Name: [(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)methyl](methyl)amine
Synonyms: [(2,2-dimethyl-3H-1-benzofuran-7-yl)methyl](methyl)amine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: ZXAQWZXMLFUWJO-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC2=C1OC(C)(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results