accessibility menu, dialog, popup

UserName

CH3BrFFF

CAS RN 86845-27-4

IUPAC Name: 4-bromo-1-methyl-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-methyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: IZFVIEYHUOUPPH-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results