accessibility menu, dialog, popup

UserName

FHOF

CAS RN 862255-56-9

IUPAC Name: 2-(2,5-difluorophenyl)ethan-1-ol
Synonyms: 2-(2,5-difluorophenyl)ethanol
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: WIOQRCFOUIBCEB-UHFFFAOYSA-N
SMILES: OCCC1=C(F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results